Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATAAGCTGCTTGAGGTTGTCATG[C/T]AGAATAGTATCCTTCACGTCACGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604406 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNA13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNA13 - G protein subunit alpha 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282425.1 | 1000 | Silent Mutation | CTA,CTG | L,L 272 | NP_001269354.1 | |
NM_006572.5 | 1000 | Silent Mutation | CTA,CTG | L,L 367 | NP_006563.2 | |
XM_011524202.1 | 1000 | Intron | XP_011522504.1 |