Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGGACCCTAAGGAAATTCTACA[C/T]TCTGTCTATGAGTGTATAGAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300474 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GK - glycerol kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000167.5 | 365 | Silent Mutation | CAC,CAT | H,H 62 | NP_000158.1 | |
NM_001128127.2 | 365 | Silent Mutation | CAC,CAT | H,H 62 | NP_001121599.1 | |
NM_001205019.1 | 365 | Silent Mutation | CAC,CAT | H,H 62 | NP_001191948.1 | |
NM_203391.3 | 365 | Silent Mutation | CAC,CAT | H,H 62 | NP_976325.1 | |
XM_005274488.4 | 365 | Intron | XP_005274545.1 | |||
XM_006724483.2 | 365 | Intron | XP_006724546.1 | |||
XM_006724484.2 | 365 | Intron | XP_006724547.1 | |||
XM_006724485.2 | 365 | Intron | XP_006724548.1 | |||
XM_006724486.3 | 365 | Intron | XP_006724549.1 | |||
XM_011545491.2 | 365 | Intron | XP_011543793.1 | |||
XM_011545492.1 | 365 | Intron | XP_011543794.1 | |||
XM_011545493.2 | 365 | Intron | XP_011543795.1 | |||
XM_011545494.2 | 365 | Intron | XP_011543796.1 | |||
XM_017029409.1 | 365 | Intron | XP_016884898.1 | |||
XM_017029410.1 | 365 | Intron | XP_016884899.1 | |||
XM_017029411.1 | 365 | Intron | XP_016884900.1 | |||
XM_017029412.1 | 365 | Intron | XP_016884901.1 |