Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTACTGCTAACCAGGGACCGCT[C/T]CTCCTGCGACCACAGCCCCGCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603293 MIM: 613992 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BNIP3 - BCL2 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004052.3 | Intron | NP_004043.3 |
PPP2R2D - protein phosphatase 2 regulatory subunit Bdelta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |