Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGACAATCTCAAGCCTGGATGCC[A/G]AAACCCCAGAATCTTCAGTGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605048 MIM: 606524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKBKE PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IKBKE - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193321.1 | Intron | NP_001180250.1 | ||||
NM_001193322.1 | Intron | NP_001180251.1 | ||||
NM_014002.3 | Intron | NP_054721.1 | ||||
XM_005273356.2 | Intron | XP_005273413.1 | ||||
XM_017002868.1 | Intron | XP_016858357.1 |
MIR6769B - microRNA 6769b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRGAP2 - SLIT-ROBO Rho GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |