Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCTCTTCTGGCGTCAGATGATC[A/G]CGGTTTCTTTTCCTTCTGCACCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DAPK1 - death associated protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288729.1 | Intron | NP_001275658.1 | ||||
NM_001288730.1 | Intron | NP_001275659.1 | ||||
NM_001288731.1 | Intron | NP_001275660.1 | ||||
NM_004938.3 | Intron | NP_004929.2 | ||||
XM_005251757.3 | Intron | XP_005251814.1 | ||||
XM_017014360.1 | Intron | XP_016869849.1 |