Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGGAGGAGGAACATGGCGGTGGC[G/T]GACCTCGCTCTCATTCCTGATGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610167 MIM: 610615 | ||||||||||||||||||||
Literature Links: |
C9orf172 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf172 - chromosome 9 open reading frame 172 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAMDC4 - MAM domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHPT1 - phosphohistidine phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135861.2 | 627 | Silent Mutation | GCG,GCT | A,A 4 | NP_001129333.1 | |
NM_001287342.1 | 627 | Silent Mutation | GCG,GCT | A,A 4 | NP_001274271.1 | |
NM_001287343.1 | 627 | Silent Mutation | GCG,GCT | A,A 4 | NP_001274272.1 | |
NM_014172.5 | 627 | Silent Mutation | GCG,GCT | A,A 4 | NP_054891.2 |
RABL6 - RAB, member RAS oncogene family-like 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |