Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCAAAGTCATCCCGCAGAGCCG[C/T]GTCGCCAAGCCGCATCAGCGCGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607336 MIM: 602913 MIM: 611713 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEST4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEST4 - bestrophin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BTBD19 - BTB domain containing 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLK3 - polo like kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004073.3 | 530 | Silent Mutation | CGC,CGT | R,R 97 | NP_004064.2 |
TCTEX1D4 - Tctex1 domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |