Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTGGAGTTTTTTGCTTGGGGA[C/T]TATTGACAGCACCCACCTTGGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD14A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD14A - major facilitator superfamily domain containing 14A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033055.2 | 400 | Silent Mutation | CTA,TTA | L,L 53 | NP_149044.2 | |
XM_005271118.1 | 400 | Silent Mutation | CTA,TTA | L,L 31 | XP_005271175.1 | |
XM_011541957.1 | 400 | Silent Mutation | CTA,TTA | L,L 53 | XP_011540259.1 | |
XM_017002084.1 | 400 | Silent Mutation | CTA,TTA | L,L 31 | XP_016857573.1 |