Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATCCCGCTGAGGAGGGCCTCTC[A/G]ATGTTCAGCCAAAGAGATGCCTAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609977 MIM: 611123 | ||||||||||||||||||||
Literature Links: |
CDCA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDCA8 - cell division cycle associated 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPHA10 - EPH receptor A10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099439.1 | 3036 | Missense Mutation | NP_001092909.1 | |||
NM_173641.2 | 3036 | Intron | NP_775912.2 | |||
XM_017001080.1 | 3036 | Missense Mutation | XP_016856569.1 | |||
XM_017001081.1 | 3036 | UTR 3 | XP_016856570.1 |