Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATCCTGAAACAGGAGCTCCAGGT[C/T]TAAGTCGGAAACACGACTGTTCCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614772 MIM: 605052 | ||||||||||||||||||||
Literature Links: |
COA6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COA6 - cytochrome c oxidase assembly factor 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARBP1 - TAR (HIV-1) RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005646.3 | 2946 | Missense Mutation | AAC,GAC | N,D 1445 | NP_005637.3 | |
XM_005273234.3 | 2946 | Missense Mutation | AAC,GAC | N,D 1444 | XP_005273291.1 | |
XM_011544263.2 | 2946 | Missense Mutation | AAC,GAC | N,D 1421 | XP_011542565.1 | |
XM_017002194.1 | 2946 | Intron | XP_016857683.1 | |||
XM_017002195.1 | 2946 | Missense Mutation | AAC,GAC | N,D 989 | XP_016857684.1 | |
XM_017002196.1 | 2946 | Missense Mutation | AAC,GAC | N,D 702 | XP_016857685.1 | |
XM_017002197.1 | 2946 | Missense Mutation | AAC,GAC | N,D 648 | XP_016857686.1 |