Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAATGAATCCTTTTTGTGTTTTG[G/T]TGGTTTTTTTTTTTTTCAGCCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604961 | ||||||||||||||||||||
Literature Links: |
PDE11A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDE11A - phosphodiesterase 11A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077196.1 | 6771 | UTR 3 | NP_001070664.1 | |||
NM_001077197.1 | 6771 | UTR 3 | NP_001070665.1 | |||
NM_001077358.1 | 6771 | Intron | NP_001070826.1 | |||
NM_016953.3 | 6771 | UTR 3 | NP_058649.3 |
TTC30A - tetratricopeptide repeat domain 30A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |