Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATACTTTGTCTTCAAATTCTT[G/T]TGGGTGGAAGAAGACCTAGTTTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604151 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2E3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBE2E3 - ubiquitin conjugating enzyme E2 E3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278554.1 | Intron | NP_001265483.1 | ||||
NM_001278555.1 | Intron | NP_001265484.1 | ||||
NM_006357.3 | Intron | NP_006348.1 | ||||
NM_182678.2 | Intron | NP_872619.1 | ||||
XM_005246244.2 | Intron | XP_005246301.1 | ||||
XM_011510492.2 | Intron | XP_011508794.1 | ||||
XM_017003166.1 | Intron | XP_016858655.1 | ||||
XM_017003167.1 | Intron | XP_016858656.1 |