Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATTTACCCAGATTTCAACCTA[A/G]GTGAATTAAAACATAGGGTTATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608165 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GULP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GULP1 - GULP, engulfment adaptor PTB domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252668.1 | Intron | NP_001239597.1 | ||||
NM_001252669.1 | Intron | NP_001239598.1 | ||||
NM_016315.3 | Intron | NP_057399.1 | ||||
XM_006712580.3 | Intron | XP_006712643.1 | ||||
XM_006712581.1 | Intron | XP_006712644.1 | ||||
XM_006712582.1 | Intron | XP_006712645.1 | ||||
XM_006712583.3 | Intron | XP_006712646.1 | ||||
XM_006712584.3 | Intron | XP_006712647.1 | ||||
XM_006712585.1 | Intron | XP_006712648.1 | ||||
XM_006712589.3 | Intron | XP_006712652.1 | ||||
XM_011511329.1 | Intron | XP_011509631.1 | ||||
XM_011511331.1 | Intron | XP_011509633.1 | ||||
XM_011511332.1 | Intron | XP_011509634.1 | ||||
XM_011511333.1 | Intron | XP_011509635.1 | ||||
XM_011511335.2 | Intron | XP_011509637.1 | ||||
XM_017004303.1 | Intron | XP_016859792.1 | ||||
XM_017004304.1 | Intron | XP_016859793.1 | ||||
XM_017004305.1 | Intron | XP_016859794.1 | ||||
XM_017004306.1 | Intron | XP_016859795.1 | ||||
XM_017004307.1 | Intron | XP_016859796.1 | ||||
XM_017004308.1 | Intron | XP_016859797.1 |
MIR561 - microRNA 561 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |