Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAAATACTGCAGTTTCTAATTAG[A/G]GTTTTGGAATAAGCTCAAGCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613817 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPATS2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPATS2L - spermatogenesis associated serine rich 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100422.1 | 2026 | Intron | NP_001093892.1 | |||
NM_001100423.1 | 2026 | Intron | NP_001093893.1 | |||
NM_001100424.1 | 2026 | Intron | NP_001093894.1 | |||
NM_001282735.1 | 2026 | Intron | NP_001269664.1 | |||
NM_001282743.1 | 2026 | Intron | NP_001269672.1 | |||
NM_001282744.1 | 2026 | Intron | NP_001269673.1 | |||
NM_015535.2 | 2026 | Intron | NP_056350.2 | |||
XM_005246456.1 | 2026 | Intron | XP_005246513.1 | |||
XM_005246457.1 | 2026 | Intron | XP_005246514.1 | |||
XM_005246458.1 | 2026 | Intron | XP_005246515.1 | |||
XM_005246459.1 | 2026 | Intron | XP_005246516.1 | |||
XM_011510938.1 | 2026 | Intron | XP_011509240.1 | |||
XM_011510939.1 | 2026 | Intron | XP_011509241.1 | |||
XM_017003783.1 | 2026 | Intron | XP_016859272.1 | |||
XM_017003784.1 | 2026 | UTR 5 | XP_016859273.1 | |||
XM_017003785.1 | 2026 | Intron | XP_016859274.1 | |||
XM_017003786.1 | 2026 | Intron | XP_016859275.1 | |||
XM_017003787.1 | 2026 | Intron | XP_016859276.1 |