Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTTTTGATGTGATTTTAGGATAT[C/T]TCAGGACAGTCTATTTGAAATGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616742 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOP58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NOP58 - NOP58 ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015934.4 | Intron | NP_057018.1 |
SNORD11B - small nucleolar RNA, C/D box 11B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD70 - small nucleolar RNA, C/D box 70 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |