Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGGGGGCCCTAGGTGCCTGCA[C/G]TTCTGCCCTCCCAACACCCAGGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600244 | ||||||||||||||||||||
Literature Links: |
PDCD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDCD1 - programmed cell death 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005018.2 | 1799 | UTR 3 | NP_005009.2 | |||
XM_006712573.2 | 1799 | Intron | XP_006712636.1 | |||
XM_017004293.1 | 1799 | UTR 3 | XP_016859782.1 |