Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCAGCTCTCTGGCCTCCATCCC[A/G]GAGACTGATCTGGGGACAGGTGGGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604282 | |||||||||||||||||||||||
Literature Links: |
H1FOO PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
H1FOO - H1 histone family member O, oocyte specific | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308262.1 | 5622 | Intron | NP_001295191.1 | |||
NM_153833.2 | 5622 | Intron | NP_722575.1 | |||
XM_017005733.1 | 5622 | Intron | XP_016861222.1 |
PLXND1 - plexin D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015103.2 | 5622 | UTR 3 | NP_055918.2 | |||
XM_011512588.1 | 5622 | Intron | XP_011510890.1 | |||
XM_011512589.1 | 5622 | UTR 3 | XP_011510891.1 | |||
XM_011512590.2 | 5622 | Intron | XP_011510892.1 | |||
XM_011512592.1 | 5622 | UTR 3 | XP_011510894.1 |