Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGCTGCATGAAACTCTGTTAGG[C/T]ACTGTACCAGCTGCTGAAACTTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600524 | ||||||||||||||||||||
Literature Links: |
RYK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RYK - receptor-like tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005861.2 | 1380 | Intron | NP_001005861.1 | |||
NM_002958.3 | 1380 | Intron | NP_002949.2 | |||
XM_017007016.1 | 1380 | Missense Mutation | TAC,TGC | Y,C 399 | XP_016862505.1 |