Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGGGGAGCGGGAGCCGCTGCT[A/C]CCGCGGATCGCCTGGGCCCACCCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612101 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PI4K2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PI4K2B - phosphatidylinositol 4-kinase type 2 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018323.3 | 221 | Silent Mutation | CTA,CTC | L,L 30 | NP_060793.2 | |
XM_005248174.1 | 221 | Silent Mutation | CTA,CTC | L,L 30 | XP_005248231.1 | |
XM_005248175.4 | 221 | Intron | XP_005248232.1 |