Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAGTCTGAGCCATCCCGTACTA[A/G]TTCATCAACTTTCCTTCCTCATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGPAT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGPAT4 - 1-acylglycerol-3-phosphate O-acyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020133.2 | 7060 | UTR 3 | NP_064518.1 | |||
XM_005267053.3 | 7060 | UTR 3 | XP_005267110.1 | |||
XM_006715514.3 | 7060 | UTR 3 | XP_006715577.1 | |||
XM_017011059.1 | 7060 | UTR 3 | XP_016866548.1 |