Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATATCTAAGATAAAAAAAAAAACC[A/C]AACCACCAAAACAACCCATTTGCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DENND2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DENND2A - DENN domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318052.1 | 3778 | UTR 3 | NP_001304981.1 | |||
NM_001318053.1 | 3778 | Intron | NP_001304982.1 | |||
NM_015689.4 | 3778 | UTR 3 | NP_056504.3 | |||
XM_005249977.3 | 3778 | UTR 3 | XP_005250034.1 | |||
XM_005249979.4 | 3778 | Intron | XP_005250036.1 | |||
XM_011516052.2 | 3778 | UTR 3 | XP_011514354.1 | |||
XM_011516053.2 | 3778 | UTR 3 | XP_011514355.1 | |||
XM_011516054.2 | 3778 | UTR 3 | XP_011514356.1 | |||
XM_017011989.1 | 3778 | UTR 3 | XP_016867478.1 |