Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCTCCTGTACCAAGTCCAGAAT[A/C]TGCATATACTGCAGAAACCACTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604281 MIM: 610821 | ||||||||||||||||||||
Literature Links: |
DBF4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBF4 - DBF4 zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318060.1 | 833 | UTR 5 | NP_001304989.1 | |||
NM_001318061.1 | 833 | UTR 5 | NP_001304990.1 | |||
NM_001318062.1 | 833 | UTR 5 | NP_001304991.1 | |||
NM_006716.3 | 833 | Missense Mutation | TAT,TCT | Y,S 110 | NP_006707.1 |
SLC25A40 - solute carrier family 25 member 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |