Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATATTTTCTGATTTCGTAGAAA[C/T]AGGTTGGCTAGCCATGGTAGTAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600917 | ||||||||||||||||||||
Literature Links: |
PPP1R3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP1R3A - protein phosphatase 1 regulatory subunit 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002711.3 | 2413 | Missense Mutation | ATT,GTT | I,V 946 | NP_002702.2 | |
XM_005250473.3 | 2413 | Missense Mutation | ATT,GTT | I,V 745 | XP_005250530.1 |