Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGAAAGGATCTCCGTTTTGCA[A/T]TGTACTTGCCTTTCCACACACTTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613301 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FEZF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FEZF1 - FEZ family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024613.3 | Intron | NP_001019784.2 | ||||
NM_001160264.2 | Intron | NP_001153736.1 | ||||
XM_005250337.3 | Intron | XP_005250394.1 | ||||
XM_011516202.2 | Intron | XP_011514504.1 |
FEZF1-AS1 - FEZF1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |