Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAATGACAGGGAGCTAGGAAGGC[A/G]AAAACACATGTAATTTTATTTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQUB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IQUB - IQ motif and ubiquitin domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA5 - NADH:ubiquinone oxidoreductase subunit A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282419.2 | 1735 | UTR 3 | NP_001269348.1 | |||
NM_001282420.2 | 1735 | UTR 3 | NP_001269349.1 | |||
NM_001282421.2 | 1735 | UTR 3 | NP_001269350.1 | |||
NM_001282422.2 | 1735 | UTR 3 | NP_001269351.1 | |||
NM_001291304.1 | 1735 | UTR 3 | NP_001278233.1 | |||
NM_005000.4 | 1735 | UTR 3 | NP_004991.1 |