Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGAAATCAAATATATAGTTAAGGT[A/G]CCATTCCTTAAAAAAATCCCATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFIB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFIB - nuclear factor I B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190737.1 | 6183 | UTR 3 | NP_001177666.1 | |||
NM_001190738.1 | 6183 | UTR 3 | NP_001177667.1 | |||
NM_001282787.1 | 6183 | UTR 3 | NP_001269716.1 | |||
NM_005596.3 | 6183 | UTR 3 | NP_005587.2 | |||
XM_005251467.2 | 6183 | UTR 3 | XP_005251524.1 | |||
XM_005251468.4 | 6183 | UTR 3 | XP_005251525.1 | |||
XM_005251469.2 | 6183 | UTR 3 | XP_005251526.1 | |||
XM_005251470.2 | 6183 | UTR 3 | XP_005251527.1 | |||
XM_005251471.3 | 6183 | UTR 3 | XP_005251528.1 | |||
XM_006716773.2 | 6183 | UTR 3 | XP_006716836.1 | |||
XM_006716774.2 | 6183 | UTR 3 | XP_006716837.1 | |||
XM_006716775.2 | 6183 | UTR 3 | XP_006716838.1 | |||
XM_017014739.1 | 6183 | UTR 3 | XP_016870228.1 | |||
XM_017014740.1 | 6183 | UTR 3 | XP_016870229.1 | |||
XM_017014741.1 | 6183 | UTR 3 | XP_016870230.1 | |||
XM_017014742.1 | 6183 | UTR 3 | XP_016870231.1 |