Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGGGAGATGGCCCCTGCCCACT[T/C]AGATCTGCTCCTTGTGCTTCACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600170 | ||||||||||||||||||||
Literature Links: |
AQP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AQP3 - aquaporin 3 (Gill blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318144.1 | 981 | Silent Mutation | CTA,CTG | L,L 220 | NP_001305073.1 | |
NM_004925.4 | 981 | Silent Mutation | TAA,TGA | *,* 293 | NP_004916.1 |