Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGCAGCTCGGTGGCTGCGGAAG[T/G]TATTGGCAATGGTGGCTACGTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601625 MIM: 606372 MIM: 601021 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ART1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ART1 - ADP-ribosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004314.2 | 4482 | Intron | NP_004305.2 | |||
XM_011520114.2 | 4482 | Intron | XP_011518416.1 | |||
XM_017017763.1 | 4482 | Intron | XP_016873252.1 |
CHRNA10 - cholinergic receptor nicotinic alpha 10 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303034.1 | 4482 | Missense Mutation | NP_001289963.1 | |||
NM_001303035.1 | 4482 | Missense Mutation | NP_001289964.1 | |||
NM_020402.3 | 4482 | Missense Mutation | NP_065135.2 | |||
XM_011520234.1 | 4482 | Missense Mutation | XP_011518536.1 |
NUP98 - nucleoporin 98 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |