Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACAGCTGCATACAGTTCAGGTA[C/G]CACCAACTCTGGTACTTGTAACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611433 | ||||||||||||||||||||
Literature Links: |
MAGOHB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAGOHB - mago homolog B, exon junction complex core component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STYK1 - serine/threonine/tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018423.2 | 1719 | Missense Mutation | CTA,GTA | L,V 400 | NP_060893.2 | |
XM_005253417.2 | 1719 | Missense Mutation | CTA,GTA | L,V 400 | XP_005253474.1 | |
XM_011520736.1 | 1719 | Missense Mutation | CTA,GTA | L,V 400 | XP_011519038.1 | |
XM_011520737.2 | 1719 | Missense Mutation | CTA,GTA | L,V 400 | XP_011519039.1 | |
XM_011520738.2 | 1719 | Missense Mutation | CTA,GTA | L,V 400 | XP_011519040.1 | |
XM_017019579.1 | 1719 | Missense Mutation | CTA,GTA | L,V 268 | XP_016875068.1 |