Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATTTGTCAGAAAGTATGAGAAT[C/T]AAATTTCACCCTTTTGTTTTCTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD13A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD13A - ankyrin repeat domain 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033121.1 | Intron | NP_149112.1 | ||||
XM_005253980.2 | Intron | XP_005254037.1 | ||||
XM_005253981.3 | Intron | XP_005254038.1 | ||||
XM_005253982.3 | Intron | XP_005254039.1 | ||||
XM_005253984.1 | Intron | XP_005254041.1 | ||||
XM_011538937.1 | Intron | XP_011537239.1 | ||||
XM_011538938.2 | Intron | XP_011537240.1 | ||||
XM_017020157.1 | Intron | XP_016875646.1 | ||||
XM_017020158.1 | Intron | XP_016875647.1 | ||||
XM_017020159.1 | Intron | XP_016875648.1 | ||||
XM_017020160.1 | Intron | XP_016875649.1 | ||||
XM_017020161.1 | Intron | XP_016875650.1 | ||||
XM_017020162.1 | Intron | XP_016875651.1 | ||||
XM_017020163.1 | Intron | XP_016875652.1 |