Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATTGTCACTTTAGGATGTTGT[A/G]TTTACTCAAAAAGCTTTCCTTATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MMAB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MMAB - methylmalonic aciduria (cobalamin deficiency) cblB type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052845.3 | 2778 | UTR 3 | NP_443077.1 | |||
XM_011538267.2 | 2778 | Intron | XP_011536569.1 | |||
XM_011538268.2 | 2778 | Intron | XP_011536570.1 | |||
XM_011538269.2 | 2778 | Intron | XP_011536571.1 |