Search Thermo Fisher Scientific
GAGTGTGCATCATTTCCTAAACTAG[C/T]TGATTGTGGAGCCCTTTTCTTGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602972 MIM: 606207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE8A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE8A - phosphodiesterase 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243137.1 | Intron | NP_001230066.1 | ||||
NM_002605.2 | Intron | NP_002596.1 | ||||
NM_173454.1 | Intron | NP_775656.1 | ||||
XM_011521668.2 | Intron | XP_011519970.1 | ||||
XM_017022308.1 | Intron | XP_016877797.1 | ||||
XM_017022309.1 | Intron | XP_016877798.1 | ||||
XM_017022310.1 | Intron | XP_016877799.1 | ||||
XM_017022311.1 | Intron | XP_016877800.1 |
SLC28A1 - solute carrier family 28 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |