Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTTAGCTGTAAAATGAAAGGAT[A/G]CTCTGAGGTCTCTACTTGTTCTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR3174 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR3174 - microRNA 3174 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF710 - zinc finger protein 710 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198526.3 | Intron | NP_940928.2 | ||||
XM_005254906.3 | Intron | XP_005254963.1 | ||||
XM_005254907.3 | Intron | XP_005254964.1 | ||||
XM_011521540.2 | Intron | XP_011519842.1 | ||||
XM_011521541.2 | Intron | XP_011519843.1 | ||||
XM_011521542.2 | Intron | XP_011519844.1 | ||||
XM_011521543.2 | Intron | XP_011519845.1 | ||||
XM_011521544.1 | Intron | XP_011519846.1 |