Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGATGAACAGAAAGAGAAGGAT[A/C]ACTATAATCTATTATTTTCTGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604686 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP13 - A-kinase anchoring protein 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270546.1 | Intron | NP_001257475.1 | ||||
NM_006738.5 | Intron | NP_006729.4 | ||||
NM_007200.4 | Intron | NP_009131.2 |
MIR7706 - microRNA 7706 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |