Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTAGACAAGTTACGTAGTCAA[A/G]CCCAGAGTTAGAGGATGGAAAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610739 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNRC6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNRC6A - trinucleotide repeat containing 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014494.2 | Intron | NP_055309.2 | ||||
XM_005255254.3 | Intron | XP_005255311.1 | ||||
XM_005255257.4 | Intron | XP_005255314.1 | ||||
XM_017023143.1 | Intron | XP_016878632.1 | ||||
XM_017023144.1 | Intron | XP_016878633.1 | ||||
XM_017023145.1 | Intron | XP_016878634.1 | ||||
XM_017023146.1 | Intron | XP_016878635.1 | ||||
XM_017023147.1 | Intron | XP_016878636.1 | ||||
XM_017023148.1 | Intron | XP_016878637.1 | ||||
XM_017023149.1 | Intron | XP_016878638.1 | ||||
XM_017023150.1 | Intron | XP_016878639.1 | ||||
XM_017023151.1 | Intron | XP_016878640.1 | ||||
XM_017023152.1 | Intron | XP_016878641.1 | ||||
XM_017023153.1 | Intron | XP_016878642.1 | ||||
XM_017023154.1 | Intron | XP_016878643.1 |