Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTTAAAGCAGGAGACGCTCAGA[A/G]ACCTGGAGAAGAAGCTGTCAGAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADGRG3 - adhesion G protein-coupled receptor G3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DRC7 - dynein regulatory complex subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289162.1 | 240 | Missense Mutation | AAC,GAC | N,D 51 | NP_001276091.1 | |
NM_001289163.1 | 240 | Missense Mutation | AAC,GAC | N,D 51 | NP_001276092.1 | |
NM_032269.5 | 240 | Missense Mutation | AAC,GAC | N,D 51 | NP_115645.4 | |
XM_011523377.1 | 240 | Missense Mutation | AAC,GAC | N,D 51 | XP_011521679.1 | |
XM_011523378.1 | 240 | Missense Mutation | AAC,GAC | N,D 51 | XP_011521680.1 |