Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAGTTTTCCTGAATGAGGTTAG[A/G]TGTCTGCTCTAAAATCCCTGTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZZEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZZEF1 - zinc finger ZZ-type and EF-hand domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015113.3 | 9073 | UTR 3 | NP_055928.3 | |||
XM_005256561.2 | 9073 | UTR 3 | XP_005256618.1 | |||
XM_011523759.2 | 9073 | Intron | XP_011522061.1 | |||
XM_017024382.1 | 9073 | Intron | XP_016879871.1 | |||
XM_017024383.1 | 9073 | Intron | XP_016879872.1 | |||
XM_017024384.1 | 9073 | Intron | XP_016879873.1 |