Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTGCTTCCTGGTGCAGAGCTCA[A/G]ATTTCAAGGTGAGCAAGAATCCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601879 | ||||||||||||||||||||
Literature Links: |
LGALS9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LGALS9 - galectin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002308.3 | 435 | Intron | NP_002299.2 | |||
NM_009587.2 | 435 | Intron | NP_033665.1 | |||
XM_006721892.2 | 435 | Intron | XP_006721955.1 | |||
XM_006721893.2 | 435 | Intron | XP_006721956.1 | |||
XM_006721894.3 | 435 | Intron | XP_006721957.1 | |||
XM_006721895.3 | 435 | Intron | XP_006721958.1 | |||
XM_011524796.1 | 435 | Missense Mutation | AAT,GAT | N,D 109 | XP_011523098.1 | |
XM_017024623.1 | 435 | Intron | XP_016880112.1 |