Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTCACTCACCCCCTCCTCCTCC[C/T]CCTCCTCCTCCTCTCCTTCCCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609966 MIM: 609293 | ||||||||||||||||||||
Literature Links: |
GGN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GGN - gametogenetin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD8 - proteasome 26S subunit, non-ATPase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPRED3 - sprouty related EVH1 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042522.2 | 476 | Missense Mutation | CCC,TCC | P,S 125 | NP_001035987.1 | |
XM_006723219.3 | 476 | Missense Mutation | CCC,TCC | P,S 125 | XP_006723282.1 | |
XM_011526978.2 | 476 | Missense Mutation | CCC,TCC | P,S 94 | XP_011525280.1 |