Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTTCCGGCGGAGGAAACTGAGG[C/T]TCAGAGAAGCTAAAAGTGTCATGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609051 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf68 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf68 - chromosome 19 open reading frame 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199341.3 | Intron | NP_955373.3 | ||||
XM_005259449.3 | Intron | XP_005259506.1 | ||||
XM_006723204.3 | Intron | XP_006723267.1 | ||||
XM_006723205.2 | Intron | XP_006723268.1 | ||||
XM_011526936.2 | Intron | XP_011525238.1 | ||||
XM_011526937.2 | Intron | XP_011525239.1 |
CARD8 - caspase recruitment domain family member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |