Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCTGGCCCAGCCCCTCCTTCTC[A/G]CCGGTGGGGGTCACCTCTGAAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604800 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX49 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX49 - DEAD-box helicase 49 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOMER3 - homer scaffolding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145721.1 | 837 | Silent Mutation | GGC,GGT | G,G 246 | NP_001139193.1 | |
NM_001145722.1 | 837 | Silent Mutation | GGC,GGT | G,G 246 | NP_001139194.1 | |
NM_001145724.1 | 837 | Silent Mutation | GGC,GGT | G,G 210 | NP_001139196.1 | |
NM_004838.3 | 837 | Silent Mutation | GGC,GGT | G,G 246 | NP_004829.3 | |
XM_006722943.1 | 837 | Silent Mutation | GGC,GGT | G,G 246 | XP_006723006.1 | |
XM_006722944.1 | 837 | Silent Mutation | GGC,GGT | G,G 246 | XP_006723007.1 |
LOC102724360 - uncharacterized LOC102724360 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |