Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGATGTTGTGAGGATTTAATGAT[C/T]TGGTGTGTGCAAAGCACAGCATAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MROH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MROH8 - maestro heat like repeat family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152503.5 | Intron | NP_689716.4 | ||||
NM_213631.2 | Intron | NP_998796.1 | ||||
NM_213632.2 | Intron | NP_998797.2 |
RPN2 - ribophorin II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135771.2 | Intron | NP_001129243.1 | ||||
NM_001324299.1 | Intron | NP_001311228.1 | ||||
NM_001324301.1 | Intron | NP_001311230.1 | ||||
NM_001324302.1 | Intron | NP_001311231.1 | ||||
NM_001324303.1 | Intron | NP_001311232.1 | ||||
NM_001324304.1 | Intron | NP_001311233.1 | ||||
NM_001324305.1 | Intron | NP_001311234.1 | ||||
NM_001324306.1 | Intron | NP_001311235.1 | ||||
NM_002951.4 | Intron | NP_002942.2 | ||||
XM_006723851.2 | Intron | XP_006723914.1 | ||||
XM_006723852.2 | Intron | XP_006723915.1 |