Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTGTGGGGTGTGCACAGCTGCC[A/G]CGCCGCCTCGTAATCAAACTCAAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600286 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PI4KA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PI4KA - phosphatidylinositol 4-kinase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_058004.3 | Intron | NP_477352.3 | ||||
XM_005261634.1 | Intron | XP_005261691.1 | ||||
XM_005261635.1 | Intron | XP_005261692.1 | ||||
XM_011530226.1 | Intron | XP_011528528.1 | ||||
XM_011530228.2 | Intron | XP_011528530.1 | ||||
XM_017028829.1 | Intron | XP_016884318.1 | ||||
XM_017028830.1 | Intron | XP_016884319.1 |