Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCAGCTCGCAGGCGCCATTCGCT[C/T]TTCAGACGCCGGAGACGTAGGAGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300092 MIM: 300044 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEX28 - testis expressed 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205201.1 | 132 | Intron | NP_001192130.1 | |||
NM_001586.2 | 132 | Intron | NP_001577.1 | |||
XM_011531116.1 | 132 | Intron | XP_011529418.1 | |||
XM_011531117.1 | 132 | Intron | XP_011529419.1 | |||
XM_011531118.2 | 132 | Intron | XP_011529420.1 |
TKTL1 - transketolase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145933.1 | 132 | UTR 5 | NP_001139405.1 | |||
NM_001145934.1 | 132 | Intron | NP_001139406.1 | |||
NM_012253.3 | 132 | UTR 5 | NP_036385.3 |