Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTAATAGCAAATCTTCTAAAAA[C/T]TGATTAAAAGGCAAAAAGGTAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM156 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM156 - transmembrane protein 156 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303228.1 | 4521 | Intron | NP_001290157.1 | |||
NM_024943.2 | 4521 | Intron | NP_079219.1 | |||
XM_005262670.3 | 4521 | Intron | XP_005262727.1 | |||
XM_011513749.2 | 4521 | Intron | XP_011512051.1 | |||
XM_011513750.2 | 4521 | Intron | XP_011512052.1 | |||
XM_011513751.2 | 4521 | UTR 3 | XP_011512053.1 | |||
XM_011513752.2 | 4521 | Intron | XP_011512054.1 | |||
XM_011513753.2 | 4521 | Intron | XP_011512055.1 | |||
XM_011513754.2 | 4521 | Intron | XP_011512056.1 | |||
XM_017008629.1 | 4521 | Intron | XP_016864118.1 |