Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCCAGGTTTCAATTTCAAGCCT[C/T]GCTTGAAGTAAAGGGATTAATAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IWS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IWS1 - IWS1, SUPT6H interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017969.2 | Intron | NP_060439.2 | ||||
XM_005263707.3 | Intron | XP_005263764.1 | ||||
XM_006712626.3 | Intron | XP_006712689.1 | ||||
XM_011511450.1 | Intron | XP_011509752.1 | ||||
XM_011511452.1 | Intron | XP_011509754.1 | ||||
XM_017004467.1 | Intron | XP_016859956.1 | ||||
XM_017004468.1 | Intron | XP_016859957.1 |