Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGGAAATTAGTCCTTTGTGATA[A/T]GAGTTGCAACTTCTTTTCCTAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNASEH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNASEH1 - ribonuclease H1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286834.1 | Intron | NP_001273763.1 | ||||
NM_001286837.1 | Intron | NP_001273766.1 | ||||
NM_002936.4 | Intron | NP_002927.2 |