Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACAACTATCTAAAGAATATAGA[C/T]CACACTCCTGCAGGGAAAGGGATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611841 MIM: 609139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL35 - mitochondrial ribosomal protein L35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REEP1 - receptor accessory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164730.1 | 1547 | UTR 3 | NP_001158202.1 | |||
NM_001164731.1 | 1547 | UTR 3 | NP_001158203.1 | |||
NM_001164732.1 | 1547 | UTR 3 | NP_001158204.1 | |||
NM_022912.2 | 1547 | UTR 3 | NP_075063.1 | |||
XM_005264502.1 | 1547 | UTR 3 | XP_005264559.1 | |||
XM_005264504.1 | 1547 | UTR 3 | XP_005264561.1 | |||
XM_011533044.1 | 1547 | UTR 3 | XP_011531346.1 | |||
XM_011533045.2 | 1547 | UTR 3 | XP_011531347.1 | |||
XM_017004725.1 | 1547 | UTR 3 | XP_016860214.1 | |||
XM_017004726.1 | 1547 | Intron | XP_016860215.1 | |||
XM_017004727.1 | 1547 | UTR 3 | XP_016860216.1 |