Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACCAATTTTCTTTTTAAACATA[A/G]CTTGTGATGGAACCCCGATTTCGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605158 | ||||||||||||||||||||
Literature Links: |
PXDN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PXDN - peroxidasin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012293.2 | 5022 | UTR 3 | NP_036425.1 | |||
XM_005264707.3 | 5022 | UTR 3 | XP_005264764.1 | |||
XM_011510396.1 | 5022 | UTR 3 | XP_011508698.1 | |||
XM_011510397.2 | 5022 | UTR 3 | XP_011508699.1 |