Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAAGTTCCTTACCATCTTTCTTT[A/T]TATTCAGTCTTCTTAACTGAGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM40 - transmembrane protein 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284406.1 | 676 | Missense Mutation | AAA,ATA | K,I 170 | NP_001271335.1 | |
NM_001284407.1 | 676 | Missense Mutation | AAA,ATA | K,I 124 | NP_001271336.1 | |
NM_001284408.1 | 676 | Missense Mutation | AAA,ATA | K,I 78 | NP_001271337.1 | |
NM_018306.3 | 676 | Missense Mutation | AAA,ATA | K,I 154 | NP_060776.2 | |
XM_011533937.2 | 676 | Missense Mutation | AAA,ATA | K,I 154 | XP_011532239.1 |